missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDCL Protein
A cDNA sequence encoding the PDCL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12988-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
5082 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTTLDD KLLGEKLQYY YSSSEDEDSD HEDKDRGRCA PASSSVPAEA ELAGEGISVN TGPKGVINDW RRFKQLETEQ REEQCREMER LIKKLSMTCR SHLDEEEEQQ KQKDLQEKIS GKMTLKEFAI MNEDQDDEEF LQQYRKQRME EMRQQLHKGP QFKQVFEISS GEGFLDMIDK EQKSIVIMVH IYEDGIPGTE AMNGCMICLA AEYPAVKFCK VKSSVIGASS QFTRNALPAL LIYKGGELIG NFVRVTDQLG DDFFAVDLEA FLQEFGLLPE KEVLVLTSVR NSATCHSEDS DLEID | |
Greater than 90% as determined by SDS-PAGE. |
PDCL Protein | |
Research Use Only | |
PDCL | |
Human | |
His | |
The PDCL solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 20% glycerol. |