missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCID2 Protein
A cDNA sequence encoding the PCID2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12975-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
PCID2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAYKCQTVIV QSFLRAFQAH KEENWALPVM YAVALDLRVF ANNADQQLVK KGKSKVGDML EKAAELLMSC FRVCASDTRA GIEDSKKWGM LFLVNQLFKI YFKINKLHLC KPLIRAIDSS NLKDDYSTAQ RVTYKYYVGR KAMFDSDFKQ AEEYLSFAFE HCHRSSQKNK RMILIYLLPV KMLLGHMPTV ELLKKYHLMQ FAEVTRAVSE GNLLLLHEAL AKHEAFFIRC GIFLILEKLK IITYRNLFKK VYLLLKTHQL SLDAFLVALK FMQVEDVDID EVQCILANLI YMGHVKGYIS HQHQKLVVSK QNPFPPLSTV C | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PCID2 protein solution (1.0 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |