missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parathyroid Hormone Protein
A DNA sequence encoding the Human Parathyroid Hormone, was expressed in the hosts and tags indicated.
105.00€ - 985.00€
Especificaciones
Número de acceso | NP_000306.1 |
---|---|
ID de gen (Entrez) | 5741 |
Nombre | Parathyroid Hormone Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Less than 1.0 EU/ug as determined by LAL method. |
Especificaciones
NP_000306.1 | |
Parathyroid Hormone Protein | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNF | |
Greater than 97% as determined by SDS-PAGE and HPLC. |
5741 | |
Research Use Only | |
PTH | |
The ED50 as determined by its ability to induce cAMP accumulation in murine MC3T3E1 cells is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |