missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parathyroid Hormone Protein
A DNA sequence encoding the Human Parathyroid Hormone, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10148-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000306.1 | |
Parathyroid Hormone Protein | |
Research Use Only | |
PTH | |
The ED50 as determined by its ability to induce cAMP accumulation in murine MC3T3E1 cells is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
5741 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNF | |
Greater than 97% as determined by SDS-PAGE and HPLC. |