missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Parathyroid Hormone Protein

A DNA sequence encoding the Human Parathyroid Hormone, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10148-ec-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922908

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Especificaciones

Especificaciones

NP_000306.1
Parathyroid Hormone Protein
Research Use Only
PTH
The ED50 as determined by its ability to induce cAMP accumulation in murine MC3T3E1 cells is less than 50 ng/ml, corresponding to a specific activity of > 2.0 M 104 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
5741
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SVSEIQLMHN LGKHLNSMER VEWLRKKLQD VHNF
Greater than 97% as determined by SDS-PAGE and HPLC.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales