missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Paired box protein Pax-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Paired box protein Pax-2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8719-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000269.3 | |
Paired box protein Pax-2 Protein | |
Research Use Only | |
PAX2 | |
Recombinant Protein | |
E. coli | |
MDMHCKADPF SAMHPGHGGV NQLGGVFVNG RPLPDVVRQR IVELAHQGVR PCDISRQLRV SHGCVSKILG RYYETGSIKP GVIGGSKPKV ATPKVVDKIA EYKRQNPTMF AWEIRDRLLA EGICDNDTVP SVSSINRIIR TKVQQPFHPT PDGAGTGVTA PGHTIVPSTA SPPVSSASND PVGSYSINGI LGIPRSNGEK RKRDEVEVYT DPAHIRGGGG LHLVWTLRDV SEGSVPNGDS QSGVDSLRKH LRADTFTQQQ LEALDRVFER PSYPDVFQAS EHIKSEQGNE YSLPALTPGL DEVKSSLSAS TNPELGSNVS GTQTYPVVTG RDMASTTLPG YPPHVPPTGQ GSYPTSTLAG MVP | |
Greater than 90% as determined by SDS-PAGE. |
5076 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |