missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OXCT1 Protein
A DNA sequence encoding the Homo sapiens (Human) OXCT1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6459-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000427.1 | |
OXCT1 Protein | |
Research Use Only | |
OXCT1 | |
Recombinant Protein | |
E. coli | |
TKFYTDPVEA VKDIPDGATV LVGGFGLCGI PENLIDALLK TGVKGLTAVS NNAGVDNFGL GLLLRSKQIK RMVSSYVGEN AEFERQYLSG ELEVELTPQG TLAERIRAGG AGVPAFYTPT GYGTLVQEGG SPIKYNKDGS VAIASKPREV REFNGQHFIL EEAITGDFAL VKAWKADRAG NVIFRKSARN FNLPMCKAAE TTVVEVEEIV DIGAFAPEDI HIPQIYVHRL IKGEKYEKRI ERLSIRKEGD GEAKSAKPGD DVRERIIKRA ALEFEDGMYA NLGIGIPLLA SNFISPNITV HLQSENGVLG LGPYPRQHEA DADLINAGKE TVTILPGASF FSSDESFAMI RGGHVDLTML GAMQVSKYGD LANWMIPGKM VKGMGGAMDL VSSAKTKVVV TMEHSAKGNA HKIMEKCTLP LTGKQCVNRI ITEKAVFDVD KKKGLTLIEL WEGLTVDDVQ KSTGCDFAVS PKLMPMQQIA N | |
Greater than 90% as determined by SDS-PAGE. |
5019 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |