missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Oncostatin M / OSM Protein
A DNA sequence encoding the Human Oncostatin M / OSM, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10361-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001306037.1 | |
Oncostatin M / OSM Protein | |
Research Use Only | |
OSM | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
5008 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
AAIGSCSKEY RVLLGQLQKQ TDLMQDTSRL LDPYIRIQGL DVPKLREHCR ERPGAFPSEE TLRGLGRRGF LQTLNATLGC VLHRLADLEQ RLPKAQDLER SGLNIEDLEK LQMARPNILG LRNNIYCMAQ LLDNSDTAEP TKAGRGASQP PTPTPASDAF QRKLEGCRFL HGYHRFMHSV GRVFSKWGES PNRSR | |
Greater than 97% as determined by SDS-PAGE and HPLC. |