missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OMP Protein
A cDNA sequence encoding the OMP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12925-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
OMP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEDRPQ QPQLDMPLVL DQGLTRQMRL RVESLKQRGE KRQDGEKLLQ PAESVYRLNF TQQQRLQFER WNVVLDKPGK VTITGTSQNW TPDLTNLMTR QLLDPTAIFW RKEDSDAIDW NEADALEFGE RLSDLAKIRK VMYFLVTFGE GVEPANLKAS VVFNQL | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
OMP protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |