missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nuclear transcription factor Y subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Nuclear transcription factor Y subunit beta, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6417-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_006157.1 | |
Nuclear transcription factor Y subunit beta Protein | |
Research Use Only | |
NFYB | |
Recombinant Protein | |
E. coli | |
MTMDGDSSTT DASQLGISAD YIGGSHYVIQ PHDDTEDSMN DHEDTNGSKE SFREQDIYLP IANVARIMKN AIPQTGKIAK DAKECVQECV SEFISFITSE ASERCHQEKR KTINGEDILF AMSTLGFDSY VEPLKLYLQK FREAMKGEKG IGGAVTATDG LSEELTEEAF TNQLPAGLIT TDGQQQNVMV YTTSYQQISG VQQIQFS | |
Greater than 80% as determined by SDS-PAGE. |
4801 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |