missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Notchless protein homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Notchless protein homolog 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8226-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001014445.1 | |
Notchless protein homolog 1 Protein | |
Research Use Only | |
NLE1 | |
Recombinant Protein | |
E. coli | |
AAAVPDEAVA RDVQRLLVQF QDEGGQLLGS PFDVPVDITP DRLQLVCNAL LAQEDPLPLA FFVHDAEIVS SLGKTLESQA VETEKVLDII YQPQAIFRVR AVTRCTSSLE GHSEAVISVA FSPTGKYLAS GSGDTTVRFW DLSTETPHFT CKGHRHWVLS ISWSPDGRKL ASGCKNGQIL LWDPSTGKQV GRTLAGHSKW ITGLSWEPLH ANPECRYVAS SSKDGSVRIW DTTAGRCERI LTGHTQSVTC LRWGGDGLLY SASQDRTIKV WRAHDGVLCR TLQGHGHWVN TMALSTDYAL RTGAFEPAEA SVNPQDLQGS LQELKERALS RYNLVRGQGP ERLVSGSDDF TLFLWSPAED KKPLTRMTGH QALINQVLFS PDSRIVASAS FDKSIKLWDG RTGKYLASLR GHVAAVYQIA WSADSRLLVS GSSDSTLKVW DVKAQKLAMD LPGHADEVYA VDWSPDGQRV ASGGKDKCLR IWRR | |
Greater than 90% as determined by SDS-PAGE. |
54475 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |