missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NECTIN3 Protein
A cDNA sequence encoding the NECTIN3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12824-1ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NECTIN3 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
GPIIVEPHVT AVWGKNVSLK CLIEVNETIT QISWEKIHGK SSQTVAVHHP QYGFSVQGEY QGRVLFKNYS LNDATITLHN IGFSDSGKYI CKAVTFPLGN AQSSTTVTVL VEPTVSLIKG PDSLIDGGNE TVAAICIAAT GKPVAHIDWE GDLGEMESTT TSFPNETATI ISQYKLFPTR FARGRRITCV VKHPALEKDI RYSFILDIQY APEVSVTGYD GNWFVGRKGV NLKCNADANP PPFKSVWSRL DGQWPDGLLA SDNTLHFVHP LTFNYSGVYI CKVTNSLGQR SDQKVIYISD PPTTTTLQPT IQWHPSTADI EDLATEPKKL PFPLSTLATI KDDTIATLEH HHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NECTIN3 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT. |