missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NANP Protein
A cDNA sequence encoding the NANP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12789-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NANP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMGLS RVRAVFFDLD NTLIDTAGAS RRGMLEVIKL LQSKYHYKEE AEIICDKVQV KLSKECFHPY NTCITDLRTS HWEEAIQETK GGAANRKLAE ECYFLWKSTR LQHMTLAEDV KAMLTELRKE VRLLLLTNGD RQTQREKIEA CACQSYFDAV VVGGEQREEK PAPSIFYYCC NLLGVQPGDC VMVGDTLETD IQGGLNAGLK ATVWINKNGI VPLKSSPVPH YMVSSVLELP ALLQSIDCKV SMST | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NANP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT and 100mM NaCl. |