missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mucosal addressin cell adhesion molecule 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Mucosal addressin cell adhesion molecule 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7559-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16069380

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
13-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_570116.2
Mucosal addressin cell adhesion molecule 1 Protein
500 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
8174
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Madcam1
Recombinant Protein
E. coli
QSLQVKPLQV EPPEPVVAVA LGASRQLTCR LACADRGASV QWRGLDTSLG AVQSDTGRSV LTVRNASLSA AGTRVCVGSC GGRTFQHTVQ LLVYAFPDQL TVSPAALVPG DPEVACTAHK VTPVDPNALS FSLLVGGQEL EGAQALGPEV QEEEEEPQGD EDVLFRVTER WRLPPLGTPV PPALYCQATM RLPGLELSHR QAIPVLHSPT SPEPPDTTSP ESPDTTSPES PDTTSQEPPD TTSPEPPDKT SPEPAPQQGS THTPRSPGST RTRRPEISQA GPTQGEVIPT GSSKPAGDQ
Videos
Documentos