missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MIF / GLIF Protein
A DNA sequence encoding the Human MIF / GLIF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10175-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002406.1 | |
MIF / GLIF Protein | |
Research Use Only | |
MIF | |
The specific activity is determined by binding rhCD74 in a functional ELISA. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
4282 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MPMFIVNTNV PRASVPDGFL SELTQQLAQA TGKPPQYIAV HVVPDQLMAF GGSSEPCALC SLHSIGKIGG AQNRSYSKLL CGLLAERLRI SPDRVYINYY DMNAANVGWN NSTFA | |
Greater than 97% as determined by SDS-PAGE and HPLC. |