missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Midkine / MDK Protein
A DNA sequence encoding the Human Midkine / MDK Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10419-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001012333.1 | |
Midkine / MDK Protein | |
Research Use Only | |
MDK | |
The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration range of 0.1-10 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH7.4, lyophilized |
4192 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
VAKKKDKVKK GGPGSECAEW AWGPCTPSSK DCGVGFREGT CGAQTQRIRC RVPCNWKKEF GADCKYKFEN WGACDGGTGT KVRQGTLKKA RYNAQCQETI RVTKPCTPKT KAKAKAKKGK GKD | |
Greater than 97% as determined by SDS-PAGE and HPLC. |