missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Melanoma-derived growth regulatory Protein
A DNA sequence encoding the Human Melanoma-derived growth regulatory protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10187-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001189482.1 | |
Melanoma-derived growth regulatory Protein | |
Research Use Only | |
MIA | |
The ED50 as determined by a cell proliferation assay using human A375 cell line is less than 5 ug/ml, corresponding to a specific activity of > 200 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 150mM NaCl, lyophilized |
8190 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPMPKLADRK LCADQECSHP ISMAVALQDY MAPDCRFLTI HRGQVVYVFS KLKGRGRLFW GGSVQGDYYG DLAARLGYFP SSIVREDQTL KPGKVDVKTD KWDFYCQ | |
Greater than 98% as determined by SDS-PAGE and HPLC. |