missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MCP-3 / CCL7 Protein
Recombinant Protein
Marca: enQuireBio™ QP5253-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
MCP-3 / CCL7 Protein | |
Research Use Only | |
CCL7 | |
Recombinant Protein | |
E. coli | |
QPVGINTSTT CCYRFINKKI PKQRLESYRR TTSSHCPREA VIFKTKLDKE ICADPTQKWV QDFMKHLDKK TQTPKL | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human MCP-3 / CCL7 induces THP-1 chemotaxis. Activity was further confirmed by measuring primary human neutrophil chemotaxis. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |