missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAPKAP1 Protein
A DNA sequence encoding the Homo sapiens (Human) MAPKAP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8011-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001006618.1 | |
MAPKAP1 Protein | |
Research Use Only | |
MAPKAP1 | |
Recombinant Protein | |
E. coli | |
AFLDNPTIIL AHIRQSHVTS DDTGMCEMVL IDHDVDLEKI HPPSMPGDSG SEIQGSNGET QGYVYAQSVD ITSSWDFGIR RRSNTAQRLE RLRKERQNQI KCKNIQWKER NSKQSAQELK SLFEKKSLKE KPPISGKQSI LSVRLEQCPL QLNNPFNEYS KFDGKGHVGT TATKKIDVYL PLHSSQDRLL PMTVVTMASA RVQDLIGLIC WQYTSEGREP KLNDNVSAYC LHIAEDDGEV DTDFPPLDSN EPIHKFGFST LALVEKYSSP GLTSKESLFV RINAAHGFSL IQVDNTKVTM KEILLKAVKR RKGSQKVSGP QYRLEKQSEP NVAVDLDSTL ESQSAWEFCL VRENSSRADG VFEEDSQIDI ATVQDMLSSH HYKSFKVSMI HRLRFTTDVQ LGISGDKVEI DPVTNQKAST KFWIKQKPIS IDSDLLCACD LAEEKSPSHA IFKLTYLSNH DYKHLYFESD AATVNEIVLK VNYILESRAS TARADYFAQK QRKLNRRTSF SFQKEKKSGQ Q | |
Greater than 90% as determined by SDS-PAGE. |
79109 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |