missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAGEA8 Protein
A cDNA sequence encoding the MAGEA8 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12626-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
4107 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLLGQKS QRYKAEEGLQ AQGEAPGLMD VQIPTAEEQK AASSSSTLIM GTLEEVTDSG SPSPPQSPEG ASSSLTVTDS TLWSQSDEGS SSNEEEGPST SPDPAHLESL FREALDEKVA ELVRFLLRKY QIKEPVTKAE MLESVIKNYK NHFPDIFSKA SECMQVIFGI DVKEVDPAGH SYILVTCLGL SYDGLLGDDQ STPKTGLLII VLGMILMEGS RAPEEAIWEA LSVMGLYDGR EHSVYWKLRK LLTQEWVQEN YLEYRQAPGS DPVRYEFLWG PRALAETSYV KVLEHVVRVN ARVRISYPSL HEEALGEEKG V | |
Greater than 85% as determined by SDS-PAGE. |
MAGEA8 Protein | |
Research Use Only | |
MAGEA8 | |
Human | |
His | |
MAGEA8 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline, 10% glycerol and 1mM DTT. |