missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LTA Protein
A DNA sequence encoding the Human LTA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1387-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000586.2 | |
LTA Protein | |
Research Use Only | |
LTA | |
The ED50 as determined by a cytotoxicity assay using murine L929 cells is less than 5 pg/ml, corresponding to a specific activity of > 2.0 M 108 IU/mg in the presence of actinomycin D. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH7.4, with 0.02 % Tween-20, lyophilized |
4049 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
LPGVGLTPSA AQTARQHPKM HLAHSTLKPA AHLIGDPSKQ NSLLWRANTD RAFLQDGFSL SNNSLLVPTS GIYFVYSQVV FSGKAYSPKA TSSPLYLAHE VQLFSSQYPF HVPLLSSQKM VYPGLQEPWL HSMYHGAAFQ LTQGDQLSTH TDGIPHLVLS PSTVFFGAFA L | |
Greater than 96% as determined by SDS-PAGE and HPLC. |