missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRRFIP1 Protein
A DNA sequence encoding the Homo sapiens (Human) LRRFIP1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6321-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Specifica
NP_001131022.1 | |
LRRFIP1 Protein | |
Research Use Only | |
LRRFIP1 | |
Recombinant Protein | |
E. coli | |
MTSPAAAQSR EIDCLSPEAQ KLAEARLAAK RAARAEAREI RMKELERQQK EVEERPEKDF TEKGSRNMPG LSAATLASLG GTSSRRGSGD TSISIDTEAS IREIKDSLAE VEEKYKKAMV SNAQLDNEKT NFMYQVDTLK DMLLELEEQL AESRRQYEEK NKEFEREKHA HSILQFQFAE VKEALKQREE MLEKHGIILN SEIATNGETS DTLNNVGYQG PTKMTKEELN ALKSTGDGTL GRASEVEVKN EIVANVGKRE ILHNTEKEQH TEDTVKDCVD IEVFPAGENT EDQKSSEDTA PFLGTLAGAT YEEQSLSTCS LDR | |
Greater than 90% as determined by SDS-PAGE. |
9208 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 3 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |