missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Long-chain-fatty-acid--CoA ligase 5 Protein
A DNA sequence encoding the Homo sapiens (Human) Long-chain-fatty-acid--CoA ligase 5, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8244-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_057318.2 | |
Long-chain-fatty-acid--CoA ligase 5 Protein | |
Research Use Only | |
ACSL5 | |
Recombinant Protein | |
E. coli | |
TRPQPVLPLL DLNNQSVGIE GGARKGVSQK NNDLTSCCFS DAKTMYEVFQ RGLAVSDNGP CLGYRKPNQP YRWLSYKQVS DRAEYLGSCL LHKGYKSSPD QFVGIFAQNR PEWIISELAC YTYSMVAVPL YDTLGPEAIV HIVNKADIAM VICDTPQKAL VLIGNVEKGF TPSLKVIILM DPFDDDLKQR GEKSGIEILS LYDAENLGKE HFRKPVPPSP EDLSVICFTS GTTGDPKGAM ITHQNIVSNA AAFLKCVEHA YEPTPDDVAI SYLPLAHMFE RIVQAVVYSC GARVGFFQGD IRLLADDMKT LKPTLFPAVP RLLNRIYDKV QNEAKTPLKK FLLKLAVSSK FKELQKGIIR HDSFWDKLIF AKIQDSLGGR VRVIVTGAAP MSTSVMTFFR AAMGCQVYEA YGQTECTGGC TFTLPGDWTS GHVGVPLACN YVKLEDVADM NYFTVNNEGE VCIKGTNVFK GYLKDPEKTQ EALDSDGWLH TGDIGRWLPN GTLKIIDRKK NIFKLAQGEY IAPEKIENIY NRSQPVLQIF VHGESLRSSL VGVVVPDTDV LPSFAAKLGV KGSFEELCQN QVVREAILED LQKIGKESGL KTFEQVKAIF LHPEPFSIEN GLLTPTLKAK RGELSKYFRT QIDSLYEHIQ D | |
Greater than 90% as determined by SDS-PAGE. |
51703 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |