missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LAG-1 Protein

A cDNA sequence encoding the LAG-1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10739-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15995018

  • 3595.00€ / 1 miligramo
Fecha estimada de envío: 03-02-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

388372; 9560
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
CCL4L1; CCL4L2
Recombinant Protein
E. coli
APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRGKQV CADPSESWVQ EYVYDLELN
LAG-1 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by a cell proliferation assay using human CCR5 transfected murine BaF3 cells is less than 2.0 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
Untagged
LAG-1 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4.
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales