missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAG-1 Protein
A cDNA sequence encoding the LAG-1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10739-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
388372; 9560 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using human CCR5 transfected murine BaF3 cells is less than 2.0 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg. | |
Human | |
Untagged | |
LAG-1 was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
LAG-1 Protein | |
Research Use Only | |
CCL4L1; CCL4L2 | |
Recombinant Protein | |
E. coli | |
APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRGKQV CADPSESWVQ EYVYDLELN | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |