Learn More
enQuireBio™ Recombinant Human Kynurenine--oxoglutarate transaminase 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Kynurenine--oxoglutarate transaminase 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7739-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001008661.1 | |
Kynurenine--oxoglutarate transaminase 3 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
56267 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CCBL | |
Recombinant Protein | |
E. coli | |
MFLAQRSLCS LSGRAKFLKT ISSSKILGFS TSAKMSLKFT NAKRIEGLDS NVWIEFTKLA ADPSVVNLGQ GFPDISPPTY VKEELSKIAA IDSLNQYTRG FGHPSLVKAL SYLYEKLYQK QIDSNKEILV TVGAYGSLFN TIQALIDEGD EVILIVPFYD CYEPMVRMAG ATPVFIPLRS KPVYGKRWSS SDWTLDPQEL ESKFNSKTKA IILNTPHNPL GKVYNREELQ VIADLCIKYD TLCISDEVYE WLVYSGNKHL KIATFPGMWE RTITIGSAGK TFSVTGWKLG WSIGPNHLIK HLQTVQQNTI YTCATPLQEA LAQAFWIDIK RMDDPECYFN SLPKELEVKR DRMVRLLESV GLKPIVPDGG YFIIADVSLL DPDLSDMKNN EPYDYKFVKW MTKHKKLSAI PVSAFCNSET KSQFEKFVRF CFIKKDSTLD AAEEIIKAWS VQKS |