missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha 2b Protein
A DNA sequence encoding the Human Interferon alpha 2b, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10405-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000596.2 | |
Interferon alpha 2b Protein | |
Research Use Only | |
IFNA2 | |
The specific activity determined by an anti-viral assay is no less than 1.0 M 108 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
3440 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
CDLPQTHSLG SRRTLMLLAQ MRKISLFSCL KDRHDFGFPQ EEFGNQFQKA ETIPVLHEMI QQIFNLFSTK DSSAAWDETL LDKFYTELYQ QLNDLEACVI QGVGVTETPL MKEDSILAVR KYFQRITLYL KEKKYSPCAW EVVRAEIMRS FSLSTNLQES LRSKE | |
Greater than 97% as determined by SDS-PAGE and HPLC. |