missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL4 / Interleukin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8557-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000580.1 | |
IL4 / Interleukin-4 Protein | |
Research Use Only | |
IL4 | |
Recombinant Protein | |
Yeast | |
HKCDITLQEI IKTLNSLTEQ KTLCTELTVT DIFAASKNTT EKETFCRAAT VLRQFYSHHE KDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE NFLERLKTIM REKYSKCSS | |
Greater than 90% as determined by SDS-PAGE. |
3565 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |