missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL21R Protein
A cDNA sequence encoding the IL21R was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12415-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
50615 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
CPDLVCYTDY LQTVICILEM WNLHPSTLTL TWQDQYEELK DEATSCSLHR SAHNATHATY TCHMDVFHFM ADDIFSVNIT DQSGNYSQEC GSFLLAESIK PAPPFNVTVT FSGQYNISWR SDYEDPAFYM LKGKLQYELQ YRNRGDPWAV SPRRKLISVD SRSVSLLPLE FRKDSSYELQ VRAGPMPGSS YQGTWSEWSD PVIFQTQSEE LKEVEHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |
IL21R Protein | |
Research Use Only | |
IL21R | |
Human | |
His | |
IL21R protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |