missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17B / IL-17B Protein
A DNA sequence encoding the Human IL17B / IL-17B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10305-ec-25ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001304916.1 | |
IL17B / IL-17B Protein | |
Research Use Only | |
IL17B | |
The ED50 as determined by inducing IL-8 secretion of human HepG2 cells is less than 1.0 ug/ml, corresponding to a specific activity of > 1000 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, with 0.1 % Tween-20 and 3 % Trehalose, lyophilized |
27190 | |
25 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MQPRSPKSKR KGQGRPGPLA PGPHQVPLDL VSRMKPYARM EEYERNIEEM VAQLRNSSEL AQRKCEVNLQ LWMSNKRSLS PWGYSINHDP SRIPVDLPEA RCLCLGCVNP FTMQEDRSMV SVPVFSQVPV RRRLCPPPPR TGPCRQRAVM ETIAVGCTCI F | |
Greater than 95% as determined by SDS-PAGE and HPLC. |