missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF1 Isoform 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha 10 / IFNA10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5005-5mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000609.1 | |
IGF1 Isoform 2 Protein | |
Research Use Only | |
IGF1 | |
Recombinant Protein | |
E. coli | |
GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA | |
Greater than 95% as determined by SDS-PAGE. |
3479 | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 10 ng/mL in the FDC-P1 cell proliferation assay. The bioactivity acceptance criteria is set at 1 .0 x 10^5 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |