missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human I-TAC / CXCL11 Protein
Recombinant Protein
Marca: enQuireBio™ QP1189-1 mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001289052.1 | |
I-TAC / CXCL11 Protein | |
Research Use Only | |
CXCL11 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6373 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
FPMFKRGRCL CIGPGVKAVK VADIEKASIM YPSNNCDKIE VIITLKENKG QRCLNPKSKQ ARLIIKKVER KNF | |
Greater than 95% as determined by SDS-PAGE. |