missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HSPA1B Protein
A cDNA sequence encoding the HSPA1B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12328-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HSPA1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS AKAAAIGIDL GTTYSCVGVF QHGKVEIIAN DQGNRTTPSY VAFTDTERLI GDAAKNQVAL NPQNTVFDAK RLIGRKFGDP VVQSDMKHWP FQVINDGDKP KVQVSYKGET KAFYPEEISS MVLTKMKEIA EAYLGYPVTN AVITVPAYFN DSQRQATKDA GVIAGLNVLR IINEPTAAAI AYGLDRTGKG ERNVLIFDLG GGTFDVSILT IDDGIFEVKA TAGDTHLGGEDFDNRLVNHF VEEFKRKHKK DISQNKRAVR RLRTACERAK RTLSSSTQAS LEIDSLFEGI DFYTSITRAR FEELCSDLFR STLEPVEKAL RDAKLDKAQI HDLVLVGGST RIPKVQKLLQ DFFNGRDLNK SINPDEAVAY GAAVQAAILM GDKSENVQDL LLLDVAPLSL GLETAGGVMT ALIKRNSTIP TKQTQIFTTY SDNQPGVLIQ VYEGERAMTK DNNLLGRFEL SGIPPAPRGVPQIEVTFDID ANGILNVTAT DKSTGKANKI TITNDKGRLS KEEIERMVQE AEKYKAEDEV QRERVSAKNA LESYAFNMKS AVEDEGLKGK ISEADKKKVL DKCQEVISWL DANTLAEKDE FEHKRKELEQ VCNPIISGLY QGAGGPGPGG FGAQGPKGGS GSGPTIEEVD |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein was filtered (0.4microm) and lyophilized from a 0.5 mg/ml solution containing 50mM phosphate buffer, 75mM NaCl, pH 7.4. |