missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homeobox protein Hox-B4 Protein
A DNA sequence encoding the Homo sapiens (Human) Homeobox protein Hox-B4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9183-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_076920.1 | |
Homeobox protein Hox-B4 Protein | |
Research Use Only | |
HOXB4 | |
Recombinant Protein | |
Yeast | |
MAMSSFLINS NYVDPKFPPC EEYSQSDYLP SDHSPGYYAG GQRRESSFQP EAGFGRRAAC TVQRYAACRD PGPPPPPPPP PPPPPPPGLS PRAPAPPPAG ALLPEPGQRC EAVSSSPPPP PCAQNPLHPS PSHSACKEPV VYPWMRKVHV STVNPNYAGG EPKRSRTAYT RQQVLELEKE FHYNRYLTRR RRVEIAHALC LSERQIKIWF QNRRMKWKKD HKLPNTKIRS GGAAGSAGGP PGRPNGGPRA L | |
Greater than 90% as determined by SDS-PAGE. |
3214 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |