missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histidine--tRNA ligase, cytoplasmic Protein
A DNA sequence encoding the Homo sapiens (Human) Histidine--tRNA ligase, cytoplasmic, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10038-ba-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001244969.1 | |
Histidine--tRNA ligase, cytoplasmic Protein | |
Research Use Only | |
HARS | |
Recombinant Protein | |
Baculovirus | |
AERAALEELV KLQGERVRGL KQQKASAELI EEEVAKLLKL KAQLGPDESK QKFVLKTPKG TRDYSPRQMA VREKVFDVII RCFKRHGAEV IDTPVFELKE TLMGKYGEDS KLIYDLKDQG GELLSLRYDL TVPFARYLAM NKLTNIKRYH IAKVYRRDNP AMTRGRYREF YQCDFDIAGN FDPMIPDAEC LKIMCEILSS LQIGDFLVKV NDRRILDGMF AICGVSDSKF RTICSSVDKL DKVSWEEVKN EMVGEKGLAP EVADRIGDYV QQHGGVSLVE QLLQDPKLSQ NKQALEGLGD LKLLFEYLTL FGIDDKISFD LSLARGLDYY TGVIYEAVLL QTPAQAGEEP LGVGSVAAGG RYDGLVGMFD PKGRKVPCVG LSIGVERIFS IVEQRLEALE EKIRTTETQV LVASAQKKLL EERLKLVSEL WDAGIKAELL YKKNPKLLNQ LQYCEEAGIP LVAIIGEQEL KDGVIKLRSV TSREEVDVRR EDLVEEIKRR TGQ | |
Greater than 90% as determined by SDS-PAGE. |
3035 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |