missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hexokinase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Hexokinase-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6164-ye-10ug
Additional Details : Peso : 0.01000kg
Specifications
NP_000179.2 | |
Hexokinase-1 Protein | |
Research Use Only | |
HK1 | |
Recombinant Protein | |
Yeast | |
ELKDDQVKKI DKYLYAMRLS DETLIDIMTR FRKEMKNGLS RDFNPTATVK MLPTFVRSIP DGSEKGDFIA LDLGGSSFRI LRVQVNHEKN QNVHMESEVY DTPENIVHGS GSQLFDHVAE CLGDFMEKRK IKDKKLPVGF TFSFPCQQSK IDEAILITWT KRFKASGVEG ADVVKLLNKA IKKRGDYDAN IVAVVNDTVG TMMTCGYDDQ HCEVGLIIGT GTNACYMEEL RHIDLVEGDE GRMCINTEWG AFGDDGSLED IRTEFDREID RGSLNPGKQL FEKMVSGMYL GELVRLILVK MAKEGLLFEG RITPELLTRG KFNTSDVSAI EKNKEGLHNA KEILTRLGVE PSDDDCVSVQ HVCTIVSFRS ANLVAATLGA ILNRLRDNKG TPRLRTTVGV DGSLYKTHPQ YSRRFHKTLR RLVPDSDVRF LLSESGSGKG AAMVTAVAYR LAEQHRQIEE TLA | |
Greater than 90% as determined by SDS-PAGE. |
3098 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |