Learn More
enQuireBio™ Recombinant Human HDAC8 / HDACL1 Protein
A DNA sequence encoding the Homo sapiens (Human) HDAC8 / HDACL1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6155-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001159890.1 | |
HDAC8 / HDACL1 Protein | |
Research Use Only | |
HDAC8 | |
Recombinant Protein | |
E. coli | |
MEEPEEPADS GQSLVPVYIY SPEYVSMCDS LAKIPKRASM VHSLIEAYAL HKQMRIVKPK VASMEEMATF HTDAYLQHLQ KVSQEGDDDH PDSIEYGLGY DCPATEGIFD YAAAIGGATI TAAQCLIDGM CKVAINWSGG WHHAKKDEAS GFCYLNDAVL GILRLRRKFE RILYVDLDLH HGDGVEDAFS FTSKVMTVSL HKFSPGFFPG TGDVSDVGLG KGRYYSVNVP IQDGIQDEKY YQICESVLKE VYQAFNPKAV VLQLGADTIA GDPMCSFNMT PVGIGKCLKY ILQWQLATLI LGGGGYNLAN TARCWTYLTG VILGKTLSSE IPDHEFFTAY GPDYVLEITP SCRPDRNEPH RIQQILNYIK GNLKHVV | |
Greater than 90% as determined by SDS-PAGE. |
55869 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |