missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPX2 Protein
A cDNA sequence encoding the GPX2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12043-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GPX2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAFIAKSFYD LSAISLDGEK VDFNTFRGRA VLIENVASLC GTTTRDFTQL NELQCRFPRR LVVLGFPCNQ FGHQENCQNE EILNSLKYVR PGGGYQPTFT LVQKCEVNGQ NEHPVFAYLK DKLPYPYDDP FSLMTDPKLI IWSPVRRSDV AWNFEKFLIG PEGEPFRRYS RTFPTINIEP DIKRLLKVAI | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GPX2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 40% glycerol, 0.15M NaCl and 1mM DTT. |