missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GPHA2 Protein
A cDNA sequence encoding the GPHA2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12038-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GPHA2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMAS QEAVIPG CHLHPFNVTV RSDRQGTCQG SHVAQACVGH CESSAFPSRY SVLVASGYRH NITSVSQCCT ISGLKKVKVQ LQCVGSRREE LEIFTARACQCDMCRLSRY | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GPHA2 filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |