missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GM-CSF / CSF2 Protein
Recombinant Protein
Marca: enQuireBio™ QP5231-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000749.2 | |
GM-CSF / CSF2 Protein | |
Research Use Only | |
CSF2 | |
Recombinant Protein | |
E. coli | |
MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE | |
Greater than 95% as determined by SDS-PAGE. |
1437 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 200 pg/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^6 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate,pH 7.5 |