missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glycerol kinase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Glycerol kinase 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6085-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_149991.2 | |
Glycerol kinase 2 Protein | |
Research Use Only | |
GK2 | |
Recombinant Protein | |
E. coli | |
AAPKTAAVGP LVGAVVQGTN STRFLVFNSK TAELLSHHKV ELTQEFPKEG WVEQDPKEIL QSVYECIART CEKLDELNID ISNIKAVGVS NQRETTVIWD KLTGEPLYNA VVWLDLRTQT TVEDLSKKIP GNSNFVKSKT GLPLSTYFSA VKLRWMLDNV RNVQKAVEEG RALFGTIDSW LIWSLTGGVN GGVHCTDVTN ASRTMLFNIH SLEWDKELCD FFEIPMDLLP NVFSSSEIYG LIKTGALEGV PISGCLGDQC AALVGQMCFQ EGQAKNTYGT GCFLLCNTGR KCVFSEHGLL TTVAYKLGRE KPAYYALEGS VAIAGAVIRW LRDNLGIIET SGDIERLAKE VGTSYGCYFV PAFSGLYAPY WEPSARGILC GLTQFTNKCH IAFAALEAVC FQTREILEAM NRDCGIPLRH LQVDGGMTNN KVLMQLQADI LHIPVIKPFM PETTALGAAM AAGAAEGVSV WSLEPQALSV LRMERFEPQI QATESEIRYA TWKKAVMKSM GWVTSQSPEG GDPSIFSSLP LGFFIVSSMV MLIGARYISG VP | |
Greater than 90% as determined by SDS-PAGE. |
2712 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |