missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galectin-7 / LGALS7 Protein
A DNA sequence encoding the Homo sapiens (Human) Galectin-7 / LGALS7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8671-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001035972.1 | |
Galectin-7 / LGALS7 Protein | |
Research Use Only | |
LGALS7 | |
Recombinant Protein | |
E. coli | |
SNVPHKSSLP EGIRPGTVLR IRGLVPPNAS RFHVNLLCGE EQGSDAALHF NPRLDTSEVV FNSKEQGSWG REERGPGVPF QRGQPFEVLI IASDDGFKAV VGDAQYHHFR HRLPLARVRL VEVGGDVQLD SVRIF | |
Greater than 90% as determined by SDS-PAGE. |
3963 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |