missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fibroblast growth factor 19 Protein
A DNA sequence encoding the Human Fibroblast growth factor 19, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10383-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005108.1 | |
Fibroblast growth factor 19 Protein | |
Research Use Only | |
FGF19 | |
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 150 ng/ml, corresponding to a specific activity of > 6.7 M 103 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
9965 | |
100 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
MRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK | |
Greater than 95% as determined by SDS-PAGE and HPLC. |