missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF21 / Fibroblast Growth Factor 21 Protein
A DNA sequence encoding the Human FGF21 / Fibroblast Growth Factor 21, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10384-ec-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_061986.1 | |
FGF21 / Fibroblast Growth Factor 21 Protein | |
Research Use Only | |
FGF21 | |
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ug/ml, corresponding to a specific activity of > 2.0 M 103 IU/mg in the presence of 5 ug/ml of rMuKlotho-beta and 10 ug/ml of heparin. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
26291 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
HPIPDSSPLL QFGGQVRQRY LYTDDAQQTE AHLEIREDGT VGGAADQSPE SLLQLKALKP GVIQILGVKT SRFLCQRPDG ALYGSLHFDP EACSFRELLL EDGYNVYQSE AHGLPLHLPG NKSPHRDPAP RGPARFLPLP GLPPALPEPP GILAPQPPDV GSSDPLSMVG PSQGRSPSYA S | |
Greater than 96% as determined by SDS-PAGE and HPLC. |