missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FAS / CD95 / APO-1 / TNFRSF6 Protein
A DNA sequence encoding the Human FAS / CD95 / APO-1 / TNFRSF6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10369-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000034.1 | |
FAS / CD95 / APO-1 / TNFRSF6 Protein | |
Research Use Only | |
FAS | |
The ED50 as determined by its ability to inhibit the cytotoxicity of Jurkat cells is between 10-15 ug/ml in the presence of 2 ng/ml of rHuFas Ligand. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
355 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
RLSSKSVNAQ VTDINSKGLE LRKTVTTVET QNLEGLHHDG QFCHKPCPPG ERKARDCTVN GDEPDCVPCQ EGKEYTDKAH FSSKCRRCRL CDEGHGLEVE INCTRTQNTK CRCKPNFFCN STVCEHCDPC TKCEHGIIKE CTLTSNTKCK EEGSRSN | |
Greater than 95% as determined by SDS-PAGE and HPLC. |