missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human F-box only protein 1

A DNA sequence encoding the Homo sapiens (Human) F-box only protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5600-ec-1mg

Additional Details : Peso : 0.01000kg

 View more versions of this product

Product Code. 15996339

  • 2450.00€ / 1 miligramo
Fecha estimada de envío: 29-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

Specifications

Specifications

NP_036300.2
F-box only protein 1
Research Use Only
FBXO2
Recombinant Protein
E. coli
MDGDGDPESV GQPEEASPEE QPEEASAEEE RPEDQQEEEA AAAAAYLDEL PEPLLLRVLA ALPAAELVQA CRLVCLRWKE LVDGAPLWLL KCQQEGLVPE GGVEEERDHW QQFYFLSKRR RNLLRNPCGE EDLEGWCDVE HGGDGWRVEE LPGDSGVEFT HDESVKKYFA SSFEWCRKAQ VIDLQAEGYW EELLDTTQPA IVVKDWYSGR SDAGCLYELT VKLLSEHENV LAEFSSGQVA VPQDSDGGGW MEISHTFTDY GPGVRFVRFE HGGQDSVYWK GWFGARVTNS SVWVEP
Greater than 80% as determined by SDS-PAGE.
26232
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers