missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human F-box only protein 1
A DNA sequence encoding the Homo sapiens (Human) F-box only protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5600-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_036300.2 | |
F-box only protein 1 | |
Research Use Only | |
FBXO2 | |
Recombinant Protein | |
E. coli | |
MDGDGDPESV GQPEEASPEE QPEEASAEEE RPEDQQEEEA AAAAAYLDEL PEPLLLRVLA ALPAAELVQA CRLVCLRWKE LVDGAPLWLL KCQQEGLVPE GGVEEERDHW QQFYFLSKRR RNLLRNPCGE EDLEGWCDVE HGGDGWRVEE LPGDSGVEFT HDESVKKYFA SSFEWCRKAQ VIDLQAEGYW EELLDTTQPA IVVKDWYSGR SDAGCLYELT VKLLSEHENV LAEFSSGQVA VPQDSDGGGW MEISHTFTDY GPGVRFVRFE HGGQDSVYWK GWFGARVTNS SVWVEP | |
Greater than 80% as determined by SDS-PAGE. |
26232 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |