missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC9 Protein
A cDNA sequence encoding the EXOSC9 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11824-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EXOSC9 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MKETPLSNCE RRFLLRAIEE KKRLDGRQTY DYRNIRISFG TDYGCCIVEL GKTRVLGQVS CELVSPKLNR ATEGILFFNL ELSQMAAPAF EPGRQSDLLV KLNRLMERCL RNSKCIDTES LCVVAGEKVW QIRVDLHLLN HDGNIIDAAS IAAIVALCHF RRPDVSVQGD EVTLYTPEER DPVPLSIHHM PICVSFAFFQ QGTYLLVDPN EREERVMDGL LVIAMNKHRE ICTIQSSGGI MLLKDQVLRC SKIAGVKVAE ITELILKALE NDQKVRKEGG KFGFAESIAN QRITAFKMEK APIDTSDVEE KAEEIIAEAE PPSEVVSTPV LWTPGTAQIG EGVENSWGDL EDSEKEDDEG GGDQAIILDG IKMDTGVEVS DIGSQDAPII LSDSEEEEMI ILEPDKNPKK IRTQTTSAKQ EKAPSKKPVK RRKKKRAAN | |
Greater than 95% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EXOSC9 is supplied in 20mM HEPES buffer pH 7.5, 0.01 mM EDTA and 0.02% SDS. |