missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC5 Protein
A cDNA sequence encoding the EXOSC5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11821-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EXOSC5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MMEEETHTDA KIRAENGTGS SPRGPGCSLR HFACEQNLLS RPDGSASFLQ GDTSVLAGVY GPAEVKVSKE IFNKATLEVI LRPKIGLPGV AEKSRERLIR NTCEAVVLGT LHPRTSITVV LQVVSDAGSL LACCLNAACM ALVDAGVPMR ALFCGVACAL DSDGTLVLDP TSKQEKEARA VLTFALDSVE RKLLMSSTKG LYSDTELQQC LAAAQAASQH VFRFYRESLQ RRYSKS | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EXOSC5 solution (0.5 mg/1ml) contains PBS buffer (pH 7.4), 200mM Nacl, 2mM DTT and 30% glycerol. |