missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC1 Protein
A cDNA sequence encoding the EXOSC1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11817-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
51013 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAPPVRY CIPGERLCNL EEGSPGSGTY TRHGYIFSSL AGCLMKSSEN GALPVVSVVR ETESQLLPDV GAIVTCKVSS INSRFAKVHI LYVGSMPLKN SFRGTIRKED VRATEKDKVE IYKSFRPGDI VLAKVISLGD AQSNYLLTTA ENELGVVVAH SESGIQMVPI SWCEMQCPKT HTKEFRKVAR VQPEFLQT | |
Greater than 90.0% as determined by SDS-PAGE. |
EXOSC1 Protein | |
Research Use Only | |
EXOSC1 | |
Human | |
His | |
EXOSC1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |